Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD11a Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: HI111]

Supplier: Biolegend
Alexa Fluor® 647 anti-human CD11a [HI111]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Capuchin monkey, Chimpanzee, Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Sheep (Ovine); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: HI111
Conjugation: Alexa Fluor® 647
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cattle (Bovine;Cow);Capuchin monkey;Chimpanzee;Cynomolgus;Dog (Canine);Horse (Equine);Rabbit (Lapine);Sheep (Ovine)

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 Tests 301218 CA301218-BL Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol CD11A
Antigen name Integrin Alpha L
Reactivity Chimpanzee, Dog, Rabbit, Macaque, Bovine, Cynomolgus Monkey, Baboon, Human, Sheep
Conjugation Alexa Fluor® 647
Clonality Monoclonal
Antibody type Primary
Host Mouse
Isotype IgG1 kappa
Flow cytometry Yes
Antigen synonyms LFA-1A, Leukocyte function-associated molecule 1 alpha chain, antigen CD11A (p180), lymphocyte function-associated antigen 1, Leukocyte adhesion glycoprotein LFA-1 alpha chain, LFA1A, CD11A, LFA-1, alpha polypeptide, CD11 antigen-like family member A, LFA-1 alpha
Gene ID
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty