Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD11c Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 3.9]

Supplier: Biolegend
301622 301620 301619
Alexa Fluor® 647 anti-human CD11c [3.9]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. <br /><strong>For test sizes</strong>, the suggested use of this reagent for immunofluorescent staining is 5 μl per million cells or 5 μl per 100 μl of whole blood.<br /><strong>For μg size</strong>, the suggested use of this reagent for immunofluorescent staining is 2 μg per 10<sup>6</sup> cells in 100 μl volume or 100 μl of whole blood.<br />It is recommended that the reagent be titrated for optimal performance for each application.<br />* Alexa Fluor® 647 has a maximum emission of 668 nm when it is excited at 633nm / 635nm.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: 3.9
Conjugation: Alexa Fluor® 647
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Chimpanzee;Cynomolgus;Rhesus;Squirrel Monkey




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 301622 CA301622-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

100 Tests 301620 CA301620-BL Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

25 tests 301619 CA301619-BL Each Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ITGAX
Antigen name Integrin Subunit Alpha X
Reactivity Cynomolgus Monkey, Macaque, Human Macaque, Cynomolgus Monkey, Human Human, Macaque, Cynomolgus Monkey
Conjugation Alexa Fluor® 647
Clonality Monoclonal
Antibody type Primary
Clone
Host Mouse
Isotype IgG1 kappa
Flow cytometry Yes
Antigen synonyms SLEB6, CD11C
Gene ID
Concentration
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty