Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD21/CD35 (CR2/CR1) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 7E9]

Supplier: Biolegend
123411 123412
APC anti-mouse CD21/CD35 (CR2/CR1) [7E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD21/CD35 (CR2/CR1)
Clonality: Monoclonal
Clone: 7E9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 123412 CA123412-BL Each (100µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

25 µg 123411 CA123411-BL Each (25µG) Retrieving

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CR1 / CR2
Antigen name Complement Component 3b/4b Receptor 1 and Complement Component 3d Receptor 2
Reactivity Mouse
Conjugation APC (Allophycocyanin)
Clonality Monoclonal
Antibody type Primary
Clone
Host Rat
Isotype IgG2a kappa
Flow cytometry Yes
Antigen synonyms CR2, C4BR, complement component 3b/4b receptor 1 (Knops blood group), C3DR, CD21, CR, CD35, C3BR, CVID7, SLEB9, KN KN, CVID7, C4BR, CR, complement component 3b/4b receptor 1 (Knops blood group), CD35, SLEB9, C3DR, C3BR, CR2, CD21
Gene ID
Concentration
Immunogen
Purification
Form
Storage temperature
Shipping temperature
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty