1,4-Anhydro-D-xylitol ≥97.0% (by HPLC)

Supplier: TCI America

A2635-100MG A2635-20MG
TCA2635-100MGEA 2472.48 CAD
TCA2635-100MG TCA2635-20MG
1,4-Anhydro-D-xylitol ≥97.0% (by HPLC)
1,4-Anhydro-D-xylitol

CAS Number: 53448-53-6
Molecular Formula: C5H10O4
Molecular Weight: 134.13
Purity/Analysis Method: >97.0% (HPLC)
Form: Clear Liquid
Boiling point (°C): 160

Formula: C₅H₁₀O₄
Boiling Pt: 160 °C
CAS Number: 53448-53-6
https://ca.vwr.com/stibo/web/std.lang.all/16/30/28931630.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR