2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane ≥98.0% (by GC)

Supplier: TCI America

B4168-5G B4168-1G
TCB4168-1GEA 259.12 CAD
TCB4168-1G TCB4168-5G
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane ≥98.0% (by GC)
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)-1,1,1,2,3,3-hexafluoro-3-(pentafluoroethoxy)propane

CAS Number: 1482416-44-3
Molecular Formula: C7BrF15O2
Molecular Weight: 480.96
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.81

Formula: C₇BrF₁₅O₂
Density: 1.81 (20 °C)
CAS Number: 1482416-44-3
https://ca.vwr.com/stibo/web/std.lang.all/54/95/28935495.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR