2-Chloro-3-hexylthiophene ≥98.0% (by GC)

Supplier: TCI America

C2613-5G
TCC2613-5GEA 388.68 CAD
TCC2613-5G
2-Chloro-3-hexylthiophene ≥98.0% (by GC)
2-Chloro-3-hexylthiophene

CAS Number: 817181-75-2
MDL Number: MFCD18413738
Molecular Formula: C10H15ClS
Molecular Weight: 202.74
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C

Formula: C₁₀H₁₅ClS
Boiling Pt: 60 °C
Density: 1.06 (20 °C)
MDL Number: MFCD18413738
CAS Number: 817181-75-2
https://ca.vwr.com/stibo/web/std.lang.all/96/59/28939659.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR