1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium (R)-2,3-Bis(tetradecanoyloxy)propyl 2,3-Dihydroxypropyl PhosphateDMPG Sodium SaltDimyristoylphosphatidylglycerol Sodium Salt

D3946-1G
TCD3946-1GEA 439.65 CAD
TCD3946-1G
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

CAS Number: 200880-40-6
MDL Number: MFCD00056649
Molecular Formula: C34H67O10P
Molecular Weight: 688.86
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids, Drug Delivery Systems (DDS) Research Reagents
Other Product Names / Synonyms: Dimyristoylphosphatidylglycerol Sodium Salt, Sodium (R)-2,3-Bis(tetradecanoyloxy)propyl 2,3-Dihydroxypropyl Phosphate, DMPG Sodium Salt

Formula: C₃₄H₆₇O₁₀P
Melting Pt: 195 °C
MDL Number: MFCD00056649
CAS Number: 200880-40-6
https://ca.vwr.com/stibo/web/std.lang.all/58/92/28985892.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR