6-Indoleboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Indole-6-boronic acid

I0803-1G
TCI0803-1GEA 562.48 CAD
TCI0803-1G
6-Indoleboronic acid (contains varying amounts of Anhydride)
6-Indoleboronic acid

CAS Number: 147621-18-9
MDL Number: MFCD03095176
Molecular Formula: C8H8BNO2
Molecular Weight: 160.97
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 200
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 6-Indolylboronic Acid

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
Melting Pt: 200 °C
MDL Number: MFCD03095176
CAS Number: 147621-18-9
https://ca.vwr.com/stibo/web/std.lang.all/26/89/28972689.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR