4'-Iodoacetanilide ≥95.0% (by GC)

Supplier: TCI America

I0846-5G I0846-1G
TCI0846-1GEA 83.74 CAD
TCI0846-5G TCI0846-1G
4'-Iodoacetanilide ≥95.0% (by GC)
4'-Iodoacetanilide

CAS Number: 622-50-4
MDL Number: MFCD00016352
Molecular Formula: C8H8INO
Molecular Weight: 261.06
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 183

https://ca.vwr.com/stibo/web/std.lang.all/27/33/28972733.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR