3-Iodocarbazole ≥98.0%
Supplier: TCI America
Synonyms:
3-Iodo-9H-carbazole
I0919-25G
I0919-5G
TCI0919-5GEA
129.56
CAD
TCI0919-5G
TCI0919-25G
3-Iodocarbazole ≥98.0%
3-Iodocarbazole
CAS Number: 16807-13-9
MDL Number: MFCD00267746
Molecular Formula: C12H8IN
Molecular Weight: 293.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 199
Lambda max.: 340 nm (CH3CN)
Formula:
C₁₂H₈IN MW: 293.11 g/mol Melting Pt: 199 °C |
MDL Number:
MFCD00267746 CAS Number: 16807-13-9 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...