1,5-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,5-Naphthaleneditriflate

N0925-5G
TCN0925-5GEA 311.26 CAD
TCN0925-5G
1,5-Naphthalenebis(trifluoromethanesulfonate) ≥98.0% (by GC)
1,5-Naphthalenebis(trifluoromethanesulfonate)

CAS Number: 152873-79-5
Molecular Formula: C12H6F6O6S2
Molecular Weight: 424.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 1,5-Naphthaleneditriflate

Formula: C₁₂H₆F₆O₆S₂
Melting Pt: 115 °C
CAS Number: 152873-79-5
https://ca.vwr.com/stibo/web/std.lang.all/01/12/28990112.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR