1,3,5-Triethynylbenzene ≥98.0% (by GC)
Supplier: TCI America
T2760-5G
T2760-1G
TCT2760-1GEA
290.74
CAD
TCT2760-1G
TCT2760-5G
1,3,5-Triethynylbenzene ≥98.0% (by GC)
1,3,5-Triethynylbenzene
CAS Number: 7567-63-7
MDL Number: MFCD00068621
Molecular Formula: C12H6
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 105
Formula:
C₁₂H₆ Melting Pt: 105 °C |
MDL Number:
MFCD00068621 CAS Number: 7567-63-7 UN: 1325 ADR: 4.1,III |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...