3-Amino-4-bromo-1H-indazole 97%

Warning

Synonyms: 4-Bromo-1H-indazol-3-amine

H33460-06 H33460-MD H33460-03 H33460-MD H33460-06 H33460-03
CAAAH33460-MDEA 299.76 CAD
CAAAH33460-MD AAH33460-03 AAH33460-MD AAH33460-06 CAAAH33460-03 CAAAH33460-06
3-Amino-4-bromo-1H-indazole 97%
3-Amino-4-bromo-1H-indazole
Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD09909580
CAS Number: 914311-50-5
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR