Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Spray Guns

https://ca.vwr.com/stibo/smallweb/std.lang.all/98/35/18989835.jpg

Spray guns are easily identified by bright coloring and feature wetted parts made of highly durable inert plastics. These sprayers are equipped with pump handles, reinforced hoses, and Viton seals for maximum chemical and acid resistance. Different spray guns may be better suited to a specific purpose with qualities that make them compatible with potent acids or bases. Rugged, stable design offers a high degree of safety for users.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Ace Glass
Description:   All-PTFE Clean-In-Place sprayer for use with either pressurized or vacuum-assisted flask washing
Product and/or packaging contains recycled/renewable content
Product is energy and/or water efficient
Catalog Number: (52848-074)
Supplier:  Bausch & Lomb
Description:   This spray pump works with 16 oz. bottles.
Catalog Number: (470106-890)
Supplier:  Ward's Science
Description:   This thumb-operated sprayer can be refilled again and again with the liquid of your choice.
Limited Availability
Catalog Number: (23001-360)
Supplier:  Terra Universal
Description:   Spray gun easily identified by natural cap color and all wetted parts are made of PFA, PTFE and polyethylene.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4