Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CAPS1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
12161R-CY5.5
CAPS1 is a 189 amino acid cytoplasmic protein that contains four EF-hand domains, which serve as calcium-binding sites, and was first identified in canine thyroid. Interestingly, CAPS1 is much less abundant in humans than in canines. Synthesis and phosphorylation of CAPS1 is upregulated by cAMP-agonists in thyrocytes. CAPS1 likely functions in the regulation of ionic transport and may be involved in cross-signaling between cAMP and Ca(+2)-phophatidylinositol cascades. In addition to thyriod, CAPS1 is expressed in brain, salivary glands and lung. Expression of CAPS1 is increased in endometrial cancer and prognosis seems to be dependent on the level of CAPS1 expression, indicating that CAPS1 may be an appropriate prognostic marker for patient survival.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPS1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 12161R-CY5.5 10225-304 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CAPS1
Antigen name Calcyphosine 1
Reactivity Mouse, Human, Rat
Conjugation Cy5.5®
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen synonyms Calcyphosin, Calcyphosine 1, CAPS, CAYP1_HUMAN, Thyroid protein p24., Calcyphosine, CAPS1
Concentration 1 μg/μl
Cross adsorption No
Purification Purified by Protein A
Form liquid
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty