Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYB5R3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
12162R-CY5.5
CYB5R3 is a 301 amino acid protein encoded by the human gene CYB5R3. CYB5R3 belongs to the flavoprotein pyridine nucleotide cytochrome reductase family and has two naturally occuring isoforms. Isoform 1 is anchored to the cytoplasmic side of the endoplasmic reticulum membrane and mitochondrion outer membrane, while isoform 2 is the soluble form found in erythrocytes. CYB5R3 is involved in the desaturation and elongation of fatty acids, cholesterol biosynthesis, drug metabolism and, in erythrocytes, methemoglobin reduction. A serine residue at position 117 seems to only be found in persons of African origin. The allele frequency is 0.23 in African Americans. It is not found in Caucasians, Asians, Indo-Aryans or Arabs. This difference seems to have no effect on the enzyme activity. Defects in CYB5R3 are the cause of hereditary methemoglobinemia (HM). There are three forms of this disease: type 1 (HM1), in which the enzyme is only deficient in erythrocytes with a mild cyanosis; type 2 (HM2), in which the enzyme is completely deficient; and type 3 (HM3), where the deficiency is seen in all blood cells. Type 2 is a severe form accompanied by mental retardation and neurological impairment.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYB5R3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 12162R-CY5.5 10225-306 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CYB5R3
Antigen name Cytochrome b5 reductase 3
Reactivity Mouse, Rat, Human
Conjugation Cy5.5®
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen synonyms diaphorase 1 (NADH), C85115, WU:AL591952.1-003, Dia1, DIA1, diaphorase 1, NADH-cytochrome b5 reductase, 2500002N19Rik, WU:AL591952.1-002, Cyb5r3, 0610016L08Rik, WU:Cyb5r3, Dia-1, WU:AL591952.1-001
Gene ID 1727
Concentration 1 μg/μl
Cross adsorption No
Purification Purified by Protein A
Form liquid
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty