Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MIB1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
12391R-CY5.5
MIB1 is a 1006 amino acid E3 ubiquitin ligase that activates the Notch ligand, Delta. MIB1 ubiquinates Delta by binding to its intracellular domain, leading to the endocytosis and eventual degradation of the Delta receptor, which, paradoxically, results in the up-regulation of receptor activity and enhances Notch signaling. MIB1 also interacts with DAPK, a protein that plays an important role in the regulation of apoptosis. Ubiquination of DAPK leads to inhibition of caspase-dependent apoptosis, therefore it is likely that overexpression of MIB1 can lead to tumor growth. Although it seems to be widely expressed at low levels, MIB1 is expressed at highest concentrations in the CNS and ovary. Both DAPK and MIB1 are overexpressed in epileptic brain tissue, suggesting that they probably cooperate as regulators of neuronal death in epilepsy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MIB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL 12391R-CY5.5 10225-678 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol MIB1
Antigen name Mindbomb E3 Ubiquitin Protein Ligase 1
Reactivity Human, Rat, Mouse
Conjugation Cy5.5®
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen synonyms MIB1, LVNC7, MIB, E3 ubiquitin-protein ligase MIB1, DAPK-interacting protein 1, KIAA1323, DIP1, DIP-1, ZZZ6, ZZANK2
Gene ID 57534
Concentration 1 μg/μl
Cross adsorption No
Purification Purified by Protein A
Form liquid
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty