Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-UBE2S Rabbit Polyclonal Antibody (Cy7®) [clone: ]

Supplier: Bioss
BS-2305R-CY7
E2 EPF is a member of the ubiquitin conjugating enzyme family. It is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme dependent manner, a characteristic property of ubiquitin carrier proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UBE2S
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL BS-2305R-CY7 10329-976 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Shipping duration cannot exceed 2 days.

restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol UBE2S
Antigen name Ubiquitin-conjugating Enzyme E2S
Reactivity Mouse, Human, Rat
Conjugation Cy7®
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Antigen synonyms EPF5, E2EPF, E2-EPF
Gene ID 27338
Concentration 1 μg/μl
Cross adsorption No
Purification Purified by Protein A
Form liquid
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty