Anti-TNFRSF9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2449R-A647
10337-828EA 1226.35 CAD
10337-828
Anti-TNFRSF9 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Receptor for TNFSF9/4-1BBL. Possibly active during T cell activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR