Anti-CCL17 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2453R-A555
10337-898EA 1226.35 CAD
10337-898
Anti-CCL17 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. TARC displays chemotactic activity for T lymphocytes, but not monocytes or granulocytes. TARC binds to chemokine receptors CCR4 and CCR8. This chemokine plays important roles in T cell development in thymus as well as in trafficking and activation of mature T cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL17
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR