Anti-CCL19 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2454R-CY5.5
10339-888EA 1226.35 CAD
10339-888
Anti-CCL19 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Strongly chemotactic for naive (L-selectinhi) CD4 T-cells and for CD8 T-cells and weakly attractive for resting B-cells and memory (L-selectinlo) CD4 T-cells. May play a role in promoting encounters between recirculating T-cells and dendritic cells and in the migration of activated B-cells into the T-zone of secondary lymphoid tissues. Binds to chemokine receptor CCR7. Binds to atypical chemokine receptor ACKR4 and mediates the recruitment of beta-arrestin (ARRB1/2) to ACKR4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL19
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR