Anti-TNFRSF14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2457R-HRP
10338-078EA 1226.35 CAD
10338-078
Anti-TNFRSF14 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
TNFRSF14 is a type I membrane protein belonging to the TNF receptor superfamily. This receptor mediates herpes virus entry into cells during infection. TNFRSF14 is able to inhibit the proliferation, activation, and cytokine production of T cells. It has an extracellular domain containing several cysteine-rich repeats and a short cytoplasmic region containing a TRAF (TNF receptor-associated factor) interaction domain. The extracellular domain of TNFRSF14 interacts with the herpes simplex virus envelope glycoprotein D. TNFRSF14 binds two cellular ligands: lymphotoxin alpha and LIGHT. LIGHT is a transmembrane protein expressed and shed from the surface of activated T cells, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes. The LIGHT:TNFRSF14 interaction controls immune response functions by cell death induction as well as cell activation. TNFRSF14 is expressed by peripheral blood T cells, B cells, monocytes and in various tissues enriched in lymphoid cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: TNFRSF14
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR