Anti-IL13RA2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2461R-CY5.5
10339-898EA 1226.35 CAD
10339-898
Anti-IL13RA2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Binds as a monomer with high affinity to interleukin-13 (IL13), but not to interleukin-4 (IL4).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR