Anti-IL13RA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-2461R-HRP
10339-904EA 1226.35 CAD
10339-904
Anti-IL13RA2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Binds as a monomer with high affinity to interleukin-13 (IL13), but not to interleukin-4 (IL4).

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: IL13RA2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR