Anti-Neuritin Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-2464R-A350
10340-086EA 1226.35 CAD
10340-086
Anti-Neuritin Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Promotes neurite outgrowth and especially branching of neuritic processes in primary hippocampal and cortical cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuritin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR