Anti-Neuritin Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-2464R-A647
10338-206EA 1225.99 CAD
10338-206
Anti-Neuritin Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Promotes neurite outgrowth and especially branching of neuritic processes in primary hippocampal and cortical cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuritin
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR