Anti-Neuritin Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2464R-CY7
10338-216EA 1226.35 CAD
10338-216
Anti-Neuritin Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Promotes neurite outgrowth and especially branching of neuritic processes in primary hippocampal and cortical cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Neuritin
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR