Anti-TNFRSF13C Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2472R-A555
10338-306EA 1226.35 CAD
10338-306
Anti-TNFRSF13C Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
B-cell receptor specific for TNFSF13B/TALL1/BAFF/BLyS. Promotes the survival of mature B-cells and the B-cell response.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF13C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR