Anti-CCL16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2481R-A555
10340-222EA 1226.35 CAD
10340-222
Anti-CCL16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
LEC (liver expressed cytokine) displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. It also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. Its expression is upregulated by IL10.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR