Anti-CCL16 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-2481R-CY5.5
10339-684EA 1226.35 CAD
10339-684
Anti-CCL16 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
LEC (liver expressed cytokine) displays chemotactic activity for lymphocytes and monocytes but not for neutrophils. It also shows a potent myelosuppressive activity and suppresses proliferation of myeloid progenitor cells. Its expression is upregulated by IL10.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL16
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR