Anti-CCL24 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-2483R-CY3
10339-704EA 1226.35 CAD
10339-704
Anti-CCL24 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Chemotactic for resting T-lymphocytes, and eosinophils. Has lower chemotactic activity for neutrophils but none for monocytes and activated lymphocytes. Is a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line. Binds to CCR3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCL24
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR