Anti-FCER2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-2490R-A555
10339-944EA 1226.35 CAD
10339-944
Anti-FCER2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Low-affinity receptor for immunoglobulin E (IgE) and CR2/CD21. Has essential roles in the regulation of IgE production and in the differentiation of B-cells (it is a B-cell-specific antigen).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD23/Fc EpsilonR II
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR