Anti-TNFRSF12A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2493R-CY7
10340-044EA 1226.35 CAD
10340-044
Anti-TNFRSF12A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor for TNFSF12/TWEAK. Weak inducer of apoptosis in some cell types. Promotes angiogenesis and the proliferation of endothelial cells. May modulate cellular adhesion to matrix proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNFRSF12A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR