Anti-CD58 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2498R-CY5.5
10340-290EA 1113.68 CAD
10340-290
Anti-CD58 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Ligand of the T-lymphocyte CD2 glycoprotein. This interaction is important in mediating thymocyte interactions with thymic epithelial cells, antigen-independent and -dependent interactions of T-lymphocytes with target cells and antigen-presenting cells and the T-lymphocyte rosetting with erythrocytes. In addition, the LFA-3/CD2 interaction may prime response by both the CD2+ and LFA-3+ cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD58
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR