Anti-CD69 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2499R-A488
10340-304EA 1114.03 CAD
10340-304
Anti-CD69 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes, natural killer (NK) cells, and platelets.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD69
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR