Anti-CD69 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2499R-CY7
10340-416EA 1226.35 CAD
10340-416
Anti-CD69 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes, natural killer (NK) cells, and platelets.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD69
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR