Anti-IRF4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

BS-2500R-A555
10340-428EA 527.87 CAD
10340-428
Anti-IRF4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Multiple myeloma oncogene-1 (MUM1) is a 50 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. to MUM1 are useful in the sub classification of lymphoid malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IRF4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR