Anti-EEF1A1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-2503R-CY5
10340-448EA 1114.03 CAD
10340-448
Anti-EEF1A1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EEF1A1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR