Anti-AIMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2509R-A350
10340-580EA 1114.03 CAD
10340-580
Anti-AIMP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Required for assembly and stability of the aminoacyl-tRNA synthase complex. Mediates ubiquitination and degradation of FUBP1, a transcriptional activator of MYC, leading to MYC down-regulation which is required for aveolar type II cell differentiation. Blocks MDM2-mediated ubiquitination and degradation of p53/TP53. Functions as a proapoptotic factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AIMP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR