Anti-CD2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-2515R-A488
10340-772EA 1226.35 CAD
10340-772
Anti-CD2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
CD2 interacts with lymphocyte function-associated antigen (LFA-3) and CD48/BCM1 to mediate adhesion between T-cells and other cell types. CD2 is implicated in the triggering of T-cells, the cytoplasmic domain is implicated in the signaling function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR