Anti-GTSE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-2516R-FITC
10340-850EA 1226.35 CAD
10340-850
Anti-GTSE1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
May be involved in p53-induced cell cycle arrest in G2/M phase by interfering with microtubule rearrangements that are required to enter mitosis. Overexpression delays G2/M phase progression.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTSE-1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR