Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-2518R-A488
10340-882EA 1225.97 CAD
10340-882
Anti-CD74 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to the endosomal/lysosomal system where the antigen processing and binding of antigenic peptides to MHC class II takes place. Serves as cell surface receptor for the cytokine MIF.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR