Anti-CD83 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-2519R-CY3
10340-912EA 1119.3 CAD
10340-912
Anti-CD83 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May play a significant role in antigen presentation or the cellular interactions that follow lymphocyte activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR