Anti-CD84 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-2520R-CY7
10340-942EA 1057.62 CAD
10340-942
Anti-CD84 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Plays a role as adhesion receptor functioning by homophilic interactions and by clustering. Recruits SH2 domain-containing proteins SH2D1A/SAP. Increases proliferative responses of activated T-cells and SH2D1A/SAP does not seen be required for this process. Homophilic interactions enhance interferon gamma/IFNG secretion in lymphocytes and induce platelet stimulation via a SH2D1A/SAP-dependent pathway. May serve as a marker for hematopoietic progenitor cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD84
Clonality: Polyclonal
Clone: 0
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR