Anti-KLRD1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-2521R-A350
10340-950EA 1226.35 CAD
10340-950
Anti-KLRD1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLRD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR