Anti-KLRD1 Rabbit Polyclonal Antibody (Biotin)

BS-2521R-BIO
10340-958EA 836.28 CAD
10340-958
Anti-KLRD1 Rabbit Polyclonal Antibody (Biotin)
Plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: KLRD1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR