Anti-ADGRE5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-2522R-A350
10340-974EA 1057.62 CAD
10340-974
Anti-ADGRE5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Receptor potentially involved in both adhesion and signaling processes early after leukocyte activation. Plays an essential role in leukocyte migration (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRE5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR