Anti-ADGRE5 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-2522R-CY7
10340-788EA 1226.35 CAD
10340-788
Anti-ADGRE5 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor potentially involved in both adhesion and signaling processes early after leukocyte activation. Plays an essential role in leukocyte migration (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRE5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR