Anti-CD99 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-2523R-CY5
10340-808EA 1226.35 CAD
10340-808
Anti-CD99 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Involved in T-cell adhesion processes. Plays a role in a late step of leukocyte extravasation helping leukocytes to overcome the endothelial basement membrane. Acts at the same site as, but independently of, PECAM1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD99
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR