Anti-CD160 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-2526R-CY5.5
10341-050EA 1114.03 CAD
10341-050
Anti-CD160 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
CD160, a 27 kDa glycoprotein, was initially identified with the monoclonal antibody BY55. Its expression is tightly associated with peripheral blood NK cells and CD8 T lymphocytes with cytolytic effector activity. The cDNA sequence of CD160 predicts a cysteine rich, glycosylphosphatidylinositol anchored protein of 181 amino acids with a single Ig-like domain weakly homologous to KIR2DL4 molecule.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD160
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR